Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 14 | 1847 | |
XM_005268209.3 | XP_005268266.1 | 14 | 1651 | ||
XM_017021765.1 | XP_016877254.1 | 14 | 1614 | ||
GenBank mRNA | AB082526.1 | - | 14 | 1783 | |
AK128693.1 | - | 14 | 2251 | ||
AL832633.2 | - | 8 | 1472 | ||
AY048580.1 | - | 14 | 1677 | ||
AY080896.1 | - | 14 | 1689 | ||
BC093881.1 | - | 14 | 1758 | 7939726 | |
BC112101.1 | - | 14 | 1758 | 8327569 |