Gene Transcripts

Gene Symbol
DIAPH2
Entrez Gene ID
1730
Gene Name
diaphanous related formin 2
Gene Aliases
DIA, DIA2, DRF2, POF, POF2
Chromosome Location
Chr. X: 96684663 - 97600598 on Build GRCh38
UniGene ID
Hs.226483
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006729.4 NP_006720.1 20 2678
NM_007309.3 NP_009293.1 20 2678
GenBank mRNA AK002029.1 - 8 950
AK291272.1 - 20 2560
BC117414.1 - 21 2345 40125965
BC143838.1 - 21 2345 9052353
U69179.1 - 2 74
Y15908.1 - 20 2632
Y15909.1 - 20 2632

View Full Product Details