Gene Transcripts

Gene Symbol
RBM18
Entrez Gene ID
92400
Gene Name
RNA binding motif protein 18
Gene Aliases
Chromosome Location
Chr. 9: 122239555 - 122264864 on Build GRCh38
UniGene ID
Hs.415842
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033117.3 NP_149108.1 4, 5 496
NR_027125.1 4, 5 496
NR_027126.1 5, 6 590
XM_006717319.1 XP_006717382.1 4, 5 452
GenBank mRNA AK057676.1 - 4, 5 472
AK299625.1 - 5, 6 566
AK310008.1 - 4, 5 453
AK310807.1 - 5, 6 532
AL389986.1 - 4, 5 424
BC008942.2 - 4, 5 443 2822634
BC019319.1 - 4, 5 443 2822634
DA308339.1 - 4, 5 496

View Full Product Details