Gene Transcripts

Gene Symbol
ZFAND4
Entrez Gene ID
93550
Gene Name
zinc finger AN1-type containing 4
Gene Aliases
ANUBL1
Chromosome Location
Chr. 10: 45614567 - 45672962 on Build GRCh38
UniGene ID
Hs.659717
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128324.2 NP_001121796.1 4, 5 580
NM_001282905.1 NP_001269834.1 5, 6 1030
NM_001282906.1 NP_001269835.1 5, 6 682
NM_174890.3 NP_777550.2 4, 5 928
NR_104256.1 4, 5 580
XM_005271842.2 XP_005271899.1 3, 4 453
XM_005271843.2 XP_005271900.1 3, 4 620
XM_017016932.1 XP_016872421.1 4, 5 672
XM_017016933.1 XP_016872422.1 4, 5 539
XM_017016934.1 XP_016872423.1 4, 5 592
XM_017016935.1 XP_016872424.1 3, 4 342
XM_017016940.1 XP_016872429.1 5, 6 1088
XM_017016941.1 XP_016872430.1 5, 6 631
XM_017016942.1 XP_016872431.1 3, 4 620
XM_017016943.1 XP_016872432.1 3, 4 896
XM_017016944.1 XP_016872433.1 3, 4 453
XM_017016946.1 XP_016872435.1 4, 5 539
GenBank mRNA AF311324.1 - 4, 5 544
AK090704.1 - 5, 6 754
AK292420.1 - 5, 6 631
AK292469.1 - 4, 5 529
AK314395.1 - 4, 5 540
BC045587.1 - 4, 5 580 5269483
BC048968.1 - 5, 6 572 5272133

View Full Product Details