Gene Transcripts

Gene Symbol
FBXO44
Entrez Gene ID
93611
Gene Name
F-box protein 44
Gene Aliases
FBG3, FBX30, FBX6A, Fbx44, Fbxo6a
Chromosome Location
Chr. 1: 11654375 - 11663327 on Build GRCh38
UniGene ID
Hs.556006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014765.1 NP_001014765.1 7 1719
NM_001304790.1 NP_001291719.1 6 1252
NM_001304791.1 NP_001291720.1 6 1458
NM_033182.5 NP_149438.2 6 1347
NM_183412.2 NP_904319.1 5 1333
NM_183413.2 NP_904320.1 5 1222
NR_130909.1 5 1088
XM_005263535.1 XP_005263592.1 6 1289
XM_005263536.4 XP_005263593.1 7 1683
XM_005263537.1 XP_005263594.1 6 1429
XM_011542435.1 XP_011540737.1 7 1751
XM_017002842.1 XP_016858331.1 7 1200
XM_017002843.1 XP_016858332.1 7 1231
GenBank mRNA AK055344.1 - 5 1333
AK316284.1 - 5 1466
AY040878.1 - 6 1303
BC007832.2 - 5 1169 4054414
CR749368.1 - 7 1708

View Full Product Details