Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001324086.1 | NP_001311015.1 | 6 | 753 | |
NM_001324410.1 | NP_001311339.1 | 6 | 654 | ||
NM_001324411.1 | NP_001311340.1 | 5 | 641 | ||
NM_001324412.1 | NP_001311341.1 | 5 | 555 | ||
NM_001324413.1 | NP_001311342.1 | 6 | 645 | ||
NM_001324414.1 | NP_001311343.1 | 6 | 701 | ||
NM_001324415.1 | NP_001311344.1 | 6 | 754 | ||
NM_058192.2 | NP_478072.1 | 6 | 744 | ||
GenBank mRNA | AL514840.3 | - | 6 | 604 | |
AL520203.3 | - | 5 | 583 | ||
BC011783.2 | - | 5 | 609 | 3937184 | |
BC012951.2 | - | 2 | 244 | ||
BC036279.2 | - | 2 | 244 | ||
BX358716.2 | - | 5 | 540 | ||
CX786721.1 | - | 6 | 704 |