Gene Transcripts

Gene Symbol
CDCA5
Entrez Gene ID
113130
Gene Name
cell division cycle associated 5
Gene Aliases
SORORIN
Chromosome Location
Chr. 11: 65061009 - 65084143 on Build GRCh38
UniGene ID
Hs.434886
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080668.3 NP_542399.1 5 454
XM_005273733.3 XP_005273790.1 5 449
XM_005273734.3 XP_005273791.1 5 449
XM_011544743.2 XP_011543045.1 5 449
XM_011544748.2 XP_011543050.1 5 449
XM_011544749.2 XP_011543051.1 5 449
XM_017017158.1 XP_016872647.1 5 449
GenBank mRNA AK093670.1 - 1 29
AK291490.1 - 5 378
AL832526.1 - 4 171
BC011000.2 - 5 362 3938081
CN332525.1 - 5 453

View Full Product Details