Gene Transcripts

Gene Symbol
MYSM1
Entrez Gene ID
114803
Gene Name
Myb like, SWIRM and MPN domains 1
Gene Aliases
2A-DUB, 2ADUB
Chromosome Location
Chr. 1: 58654739 - 58700091 on Build GRCh38
UniGene ID
Hs.744921
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085487.2 NP_001078956.1 7 499
XM_006710314.3 XP_006710377.1 7 515
XM_011540573.2 XP_011538875.1 8 479
XM_011540574.2 XP_011538876.1 7 747
XM_017000195.1 XP_016855684.1 8 551
XM_017000196.1 XP_016855685.1 5 401
XM_017000197.1 XP_016855686.1 3 179
GenBank mRNA AB067502.1 - 3 525
AK126835.1 - 1 48
BX648168.1 - 4 235

View Full Product Details