Gene Transcripts

Gene Symbol
SMIM19
Entrez Gene ID
114926
Gene Name
small integral membrane protein 19
Gene Aliases
C8orf40
Chromosome Location
Chr. 8: 42541155 - 42552997 on Build GRCh38
UniGene ID
Hs.655320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135674.1 NP_001129146.1 2 871
NM_001135675.1 NP_001129147.1 2 479
NM_001135676.1 NP_001129148.1 2 164
NM_138436.3 NP_612445.2 2 250
XM_005273398.4 XP_005273455.1 2 785
GenBank mRNA AK311932.1 - 2 225
BC013035.2 - 2 212 3535608
BI463786.1 - 2 169
BI549696.1 - 2 484
DA702519.1 - 2 250

View Full Product Details