Gene Transcripts
- Gene Symbol
- DNASE1
- Entrez Gene ID
- 1773
- Gene Name
- deoxyribonuclease I
- Gene Aliases
- DNL1, DRNI
- Chromosome Location
- Chr. 16: 3611705 - 3665472 on Build GRCh38
- UniGene ID
- Hs.629638
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_005223.3 |
NP_005214.2
|
7, 8 |
2819 |
|
XM_005255148.3 |
XP_005255205.1
|
10, 11 |
1730 |
|
XM_006720854.3 |
XP_006720917.1
|
9, 10 |
1642 |
|
XM_011522393.2 |
XP_011520695.1
|
10, 11 |
1729 |
|
XM_017022992.1 |
XP_016878481.1
|
10, 11 |
1731 |
|
XM_017022993.1 |
XP_016878482.1
|
11, 12 |
2336 |
|
XM_017022994.1 |
XP_016878483.1
|
12, 13 |
2502 |
|
XM_017022995.1 |
XP_016878484.1
|
10, 11 |
1536 |
|
XM_017022996.1 |
XP_016878485.1
|
10, 11 |
1482 |
|
XM_017022997.1 |
XP_016878486.1
|
11, 12 |
2150 |
|
XM_017022998.1 |
XP_016878487.1
|
9, 10 |
2435 |
|
XM_017022999.1 |
XP_016878488.1
|
8, 9 |
1473 |
|
XM_017023000.1 |
XP_016878489.1
|
6, 7 |
1271 |
|
XM_017023001.1 |
XP_016878490.1
|
9, 10 |
1644 |
|
XM_017023002.1 |
XP_016878491.1
|
6, 7 |
1549 |
|
XM_017023003.1 |
XP_016878492.1
|
6, 7 |
1525 |
|
XM_017023004.1 |
XP_016878493.1
|
8, 9 |
1533 |
|
XM_017023005.1 |
XP_016878494.1
|
9, 10 |
2138 |
|
XM_017023006.1 |
XP_016878495.1
|
4, 5 |
454 |
|
XM_017023007.1 |
XP_016878496.1
|
9, 10 |
1617 |
|
GenBank mRNA |
AB188151.1 |
- |
8, 9 |
898 |
|
AB188152.1 |
- |
7, 8 |
952 |
|
AJ298844.1 |
- |
7, 8 |
1774 |
|
AK300914.1 |
- |
5, 6 |
721 |
|
BC029437.1 |
- |
7, 8 |
840 |
4594247 |
BX648257.1 |
- |
5, 6 |
728 |
|
M55983.1 |
- |
7, 8 |
871 |
|
View Full Product Details