Gene Transcripts

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
Chromosome Location
Chr. 21: 26466209 - 26573404 on Build GRCh38
UniGene ID
Hs.37445
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320768.1 NP_001307697.1 3 667
NM_052954.3 NP_443186.2 3 667
NR_135472.1 4 834
XM_011529450.2 XP_011527752.1 3 547
GenBank mRNA AF401639.1 - 3 272
AK223576.1 - 3 384
AL833200.1 - 3 478
AY061853.1 - 3 212
BC036761.1 - 3 455 5588436
JF693912.1 - 3 215

View Full Product Details