Gene Transcripts

Gene Symbol
MRGPRX4
Entrez Gene ID
117196
Gene Name
MAS related GPR family member X4
Gene Aliases
GPCR, MRGX4, SNSR6
Chromosome Location
Chr. 11: 18172837 - 18174280 on Build GRCh38
UniGene ID
Hs.632138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054032.3 NP_473373.2 1 933
GenBank mRNA BC095509.1 - 1 943
BC107099.1 - 1 513 40015896
BC107100.1 - 1 513 40015904

View Full Product Details