Gene Transcripts

Gene Symbol
DRG2
Entrez Gene ID
1819
Gene Name
developmentally regulated GTP binding protein 2
Gene Aliases
Chromosome Location
Chr. 17: 18087867 - 18107985 on Build GRCh38
UniGene ID
Hs.78582
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001388.4 NP_001379.1 7 727
XM_005256499.2 XP_005256556.1 7 709
XM_005256500.2 XP_005256557.1 7 704
XM_011523704.1 XP_011522006.1 7 709
XM_011523705.1 XP_011522007.1 7 709
XM_011523706.1 XP_011522008.1 7 638
XM_011523707.1 XP_011522009.1 7 684
XM_017024296.1 XP_016879785.1 7 638
XM_017024297.1 XP_016879786.1 7 684
GenBank mRNA AB209340.1 - 3 4088
AK297569.1 - 6 1256
AK313362.1 - 7 646
BC000493.2 - 7 625 2820542
BT006976.1 - 7 570
X80754.1 - 7 617

View Full Product Details