Gene Transcripts
- Gene Symbol
- DRG2
- Entrez Gene ID
- 1819
- Gene Name
- developmentally regulated GTP binding protein 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 18087867 - 18107985 on Build GRCh38
- UniGene ID
- Hs.78582
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001388.4 |
NP_001379.1
|
8, 9 |
887 |
|
XM_005256499.2 |
XP_005256556.1
|
8, 9 |
869 |
|
XM_005256500.2 |
XP_005256557.1
|
8, 9 |
864 |
|
XM_011523704.1 |
XP_011522006.1
|
8, 9 |
869 |
|
XM_011523705.1 |
XP_011522007.1
|
8, 9 |
869 |
|
XM_011523706.1 |
XP_011522008.1
|
8, 9 |
798 |
|
XM_011523707.1 |
XP_011522009.1
|
8, 9 |
844 |
|
XM_017024296.1 |
XP_016879785.1
|
8, 9 |
798 |
|
XM_017024297.1 |
XP_016879786.1
|
8, 9 |
844 |
|
GenBank mRNA |
AB209340.1 |
- |
4, 5 |
4248 |
|
AK297569.1 |
- |
7, 8 |
1416 |
|
AK313362.1 |
- |
8, 9 |
806 |
|
BC000493.2 |
- |
8, 9 |
785 |
2820542 |
BT006976.1 |
- |
8, 9 |
730 |
|
X80754.1 |
- |
8, 9 |
777 |
|
View Full Product Details