Gene Transcripts

Gene Symbol
RPTN
Entrez Gene ID
126638
Gene Name
repetin
Gene Aliases
Chromosome Location
Chr. 1: 152153595 - 152159228 on Build GRCh38
UniGene ID
Hs.376144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122965.1 NP_001116437.1 3 1892
GenBank mRNA AK096436.1 - 1 802
AY396742.1 - 3 1892

View Full Product Details