Gene Transcripts
- Gene Symbol
- TDRD10
- Entrez Gene ID
- 126668
- Gene Name
- tudor domain containing 10
- Gene Aliases
- Chromosome Location
- Chr. 1: 154502219 - 154548147 on Build GRCh38
- UniGene ID
- Hs.387671
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001098475.1 |
NP_001091945.1
|
7, 8 |
1250 |
|
NM_182499.3 |
NP_872305.3
|
7, 8 |
1250 |
|
XM_006711156.2 |
XP_006711219.1
|
2, 3 |
880 |
|
XM_011509152.1 |
XP_011507454.1
|
8, 9 |
639 |
|
XM_011509153.1 |
XP_011507455.1
|
8, 9 |
639 |
|
XM_011509154.1 |
XP_011507456.1
|
8, 9 |
597 |
|
XM_011509155.1 |
XP_011507457.1
|
8, 9 |
639 |
|
XM_011509156.1 |
XP_011507458.1
|
7, 8 |
564 |
|
XM_011509157.1 |
XP_011507459.1
|
7, 8 |
564 |
|
XM_011509158.2 |
XP_011507460.1
|
3, 4 |
354 |
|
XM_011509159.2 |
XP_011507461.1
|
3, 4 |
345 |
|
XM_011509160.2 |
XP_011507462.1
|
3, 4 |
390 |
|
XM_011509161.1 |
XP_011507463.1
|
3, 4 |
358 |
|
XM_017000255.1 |
XP_016855744.1
|
2, 3 |
284 |
|
GenBank mRNA |
AK302144.1 |
- |
7, 8 |
497 |
|
AL713777.1 |
- |
2, 3 |
156 |
|
AY358195.1 |
- |
1, 2 |
19 |
|
BC044864.1 |
- |
7, 8 |
1249 |
4836747 |
BC100895.1 |
- |
2, 3 |
128 |
|
BC100896.1 |
- |
2, 3 |
128 |
|
BC100897.2 |
- |
2, 3 |
128 |
|
BC100898.2 |
- |
2, 3 |
128 |
|
View Full Product Details