Gene Transcripts

Gene Symbol
ASB17
Entrez Gene ID
127247
Gene Name
ankyrin repeat and SOCS box containing 17
Gene Aliases
Asb-17
Chromosome Location
Chr. 1: 75918873 - 75937625 on Build GRCh38
UniGene ID
Hs.125423
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080868.2 NP_543144.1 2 726
NR_026546.1 2 664
XM_006710345.3 XP_006710408.1 3 558
XM_006710346.3 XP_006710409.1 3 485
GenBank mRNA AF403035.1 - 2 586
AF429968.1 - 2 703
AK098606.1 - 2 726
BC022517.1 - 2 597
BC036573.2 - 2 704 5297837

View Full Product Details