Gene Transcripts

Gene Symbol
OR6K6
Entrez Gene ID
128371
Gene Name
olfactory receptor family 6 subfamily K member 6
Gene Aliases
OR1-21
Chromosome Location
Chr. 1: 158754816 - 158755847 on Build GRCh38
UniGene ID
Hs.553591
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005184.1 NP_001005184.1 1 223
GenBank mRNA BC140743.1 - 1 319

View Full Product Details