Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138801.2 | NP_620156.1 | 6 | 1123 | |
GenBank mRNA | AK291489.1 | - | 6 | 931 | |
BC014916.1 | - | 6 | 905 | 3920640 | |
BC019263.1 | - | 6 | 936 | 3502667 | |
EU794611.1 | - | 6 | 905 |