Gene Transcripts

Gene Symbol
DTYMK
Entrez Gene ID
1841
Gene Name
deoxythymidylate kinase
Gene Aliases
CDC8, PP3731, TMPK, TYMK
Chromosome Location
Chr. 2: 241675742 - 241686968 on Build GRCh38
UniGene ID
Hs.471873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165031.1 NP_001158503.1 2 327
NM_001320902.1 NP_001307831.1 2 327
NM_001320903.1 NP_001307832.1 2 327
NM_001320904.1 NP_001307833.1 2 327
NM_001320905.1 NP_001307834.1 2 327
NM_012145.3 NP_036277.2 2 327
NR_033255.1 2 327
NR_135492.1 2 327
GenBank mRNA AA427388.1 - 2 68
AK223434.1 - 2 174
BC001827.2 - 2 167 3533327
BC147006.1 - 2 266 9056934
BC147010.1 - 2 266 9056938
BC171902.1 - 2 266 9054556
BC171903.1 - 2 266
BG765702.1 - 2 73
BM552707.1 - 2 313
CN356277.1 - 2 163
CR541986.1 - 2 142
CR542015.1 - 2 142
CX166352.1 - 2 327
L16991.1 - 2 184
X54729.1 - 2 166

View Full Product Details