Gene Transcripts
- Gene Symbol
- DTYMK
- Entrez Gene ID
- 1841
- Gene Name
- deoxythymidylate kinase
- Gene Aliases
- CDC8, PP3731, TMPK, TYMK
- Chromosome Location
- Chr. 2: 241675742 - 241686968 on Build GRCh38
- UniGene ID
- Hs.471873
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001165031.1 |
NP_001158503.1
|
4, 5 |
649 |
|
NM_001320902.1 |
NP_001307831.1
|
3, 4 |
592 |
|
NM_001320903.1 |
NP_001307832.1
|
3, 4 |
558 |
|
NM_001320904.1 |
NP_001307833.1
|
4, 5 |
664 |
|
NM_001320905.1 |
NP_001307834.1
|
5, 6 |
838 |
|
NM_012145.3 |
NP_036277.2
|
4, 5 |
721 |
|
NR_033255.1 |
|
3, 4 |
630 |
|
NR_135492.1 |
|
4, 5 |
683 |
|
GenBank mRNA |
AK223434.1 |
- |
4, 5 |
568 |
|
BC001827.2 |
- |
4, 5 |
561 |
3533327 |
BC147006.1 |
- |
4, 5 |
660 |
9056934 |
BC147010.1 |
- |
4, 5 |
660 |
9056938 |
BC171894.1 |
- |
4, 5 |
656 |
|
BC171902.1 |
- |
4, 5 |
588 |
9054556 |
BC171903.1 |
- |
3, 4 |
569 |
|
BG765702.1 |
- |
3, 4 |
376 |
|
BM552707.1 |
- |
4, 5 |
707 |
|
CN356277.1 |
- |
3, 4 |
428 |
|
CR541986.1 |
- |
4, 5 |
536 |
|
CR542015.1 |
- |
4, 5 |
536 |
|
CX166352.1 |
- |
4, 5 |
721 |
|
L16991.1 |
- |
4, 5 |
578 |
|
X54729.1 |
- |
4, 5 |
560 |
|
View Full Product Details