Gene Transcripts

Gene Symbol
DUSP5
Entrez Gene ID
1847
Gene Name
dual specificity phosphatase 5
Gene Aliases
DUSP, HVH3
Chromosome Location
Chr. 10: 110497867 - 110511544 on Build GRCh38
UniGene ID
Hs.2128
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004419.3 NP_004410.3 3 967
GenBank mRNA AB209823.1 - 3 913
BC062545.1 - 3 884 5808876
U15932.2 - 3 919
U16996.1 - 3 758

View Full Product Details