Gene Transcripts

Gene Symbol
ACOT12
Entrez Gene ID
134526
Gene Name
acyl-CoA thioesterase 12
Gene Aliases
CACH-1, Cach, STARD15, THEAL
Chromosome Location
Chr. 5: 81309045 - 81394174 on Build GRCh38
UniGene ID
Hs.591756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130767.2 NP_570123.1 13 1417
XM_006714531.2 XP_006714594.1 12 1298
XM_017009045.1 XP_016864534.1 13 1382
XM_017009046.1 XP_016864535.1 12 1298
XM_017009047.1 XP_016864536.1 12 1320
XM_017009048.1 XP_016864537.1 9 1094
GenBank mRNA AB078619.1 - 13 1385
AK122960.1 - 13 1382
BC075010.2 - 13 1383 30915616
BC075011.2 - 13 1378 30915403

View Full Product Details