Gene Transcripts

Gene Symbol
CLVS2
Entrez Gene ID
134829
Gene Name
clavesin 2
Gene Aliases
C6orf212, C6orf213, RLBP1L2, bA160A10.4
Chromosome Location
Chr. 6: 122993693 - 123081282 on Build GRCh38
UniGene ID
Hs.486361
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010852.3 NP_001010852.2 5 2039
XM_017010266.1 XP_016865755.1 5 3741
GenBank mRNA AK095527.1 - 5 2043
AK295158.1 - 5 501
BC101376.1 - 5 1573 40025886
BC101377.2 - 5 1575 40025889

View Full Product Details