Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001139456.1 | NP_001132928.1 | 12 | 1243 | |
NM_174959.3 | NP_777619.1 | 8 | 860 | ||
XM_005250143.3 | XP_005250200.1 | 13 | 1647 | ||
XM_011515797.2 | XP_011514099.1 | 9 | 934 | ||
XM_017011746.1 | XP_016867235.1 | 10 | 1049 | ||
XM_017011747.1 | XP_016867236.1 | 9 | 874 | ||
XM_017011748.1 | XP_016867237.1 | 9 | 879 | ||
GenBank mRNA | BC036796.1 | - | 8 | 860 | |
BX349158.2 | - | 3 | 290 | ||
BX648789.1 | - | 8 | 4480 |