Gene Transcripts

Gene Symbol
OR10A5
Entrez Gene ID
144124
Gene Name
olfactory receptor family 10 subfamily A member 5
Gene Aliases
JCG6, OR10A1, OR11-403
Chromosome Location
Chr. 11: 6845683 - 6846636 on Build GRCh38
UniGene ID
Hs.447478
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178168.1 NP_835462.1 1 97
GenBank mRNA AF324499.1 - 1 97
BC093651.1 - 1 128 7939496
BC093653.1 - 1 128 7939498

View Full Product Details