Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006931.2 | NP_008862.1 | 10 | 1577 | |
GenBank mRNA | AB209607.1 | - | 9 | 2056 | |
AK074861.1 | - | 2 | 218 | ||
AK304509.1 | - | 10 | 1280 | ||
AK312386.1 | - | 10 | 1468 | ||
AY034634.1 | - | 4 | 414 | ||
BC039196.1 | - | 10 | 1452 | 4396508 | |
CR457413.1 | - | 9 | 1315 | ||
M20681.1 | - | 10 | 1557 |