Gene Transcripts

Gene Symbol
OVOS
Entrez Gene ID
408186
Gene Name
ovostatin
Gene Aliases
Chromosome Location
Chr. 12: 9517803 - 9573287 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020287.1 XP_016875776.1 7 672

Gene Symbol
OVOS2
Entrez Gene ID
144203
Gene Name
ovostatin 2
Gene Aliases
Chromosome Location
Chr. 12: 31114623 - 31206154 on Build GRCh38
UniGene ID
Hs.524331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020288.1 XP_016875777.1 8 876

View Full Product Details