Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256283.1 | NP_001243212.1 | 3 | 716 | |
NM_001957.3 | NP_001948.1 | 4 | 1206 | ||
GenBank mRNA | AF014826.1 | - | 2 | 396 | |
AK304451.1 | - | 3 | 488 | ||
AK312812.1 | - | 4 | 1026 | ||
AK315931.1 | - | 3 | 701 | ||
AY275462.1 | - | 3 | 676 | ||
BC022511.1 | - | 4 | 1026 | 4812050 | |
D90348.1 | - | 4 | 1156 | ||
L06622.1 | - | 3 | 682 | ||
S45956.1 | - | 4 | 897 | ||
S57498.1 | - | 4 | 1160 | ||
S63938.1 | - | 3 | 749 | ||
S67127.1 | - | 3 | 813 | ||
S81539.1 | - | 3 | 676 | ||
X61950.1 | - | 4 | 1160 |