Gene Transcripts

Gene Symbol
TTC7B
Entrez Gene ID
145567
Gene Name
tetratricopeptide repeat domain 7B
Gene Aliases
TTC7L1, c14_5685
Chromosome Location
Chr. 14: 90540588 - 90816479 on Build GRCh38
UniGene ID
Hs.655697
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010854.1 NP_001010854.1 15, 16 1867
NM_001320421.1 NP_001307350.1 15, 16 1863
XM_005267367.1 XP_005267424.1 11, 12 1178
XM_011536497.1 XP_011534799.1 15, 16 1925
XM_017021044.1 XP_016876533.1 15, 16 1831
XM_017021045.1 XP_016876534.1 15, 16 1831
XM_017021046.1 XP_016876535.1 15, 16 1925
XM_017021047.1 XP_016876536.1 14, 15 1542
GenBank mRNA AK126571.1 - 6, 7 600
AL832848.1 - 14, 15 1545
BC035865.2 - 9, 10 968
BC048270.2 - 9, 10 968
BX247966.1 - 1, 2 155
BX248275.1 - 11, 12 1159

View Full Product Details