Gene Transcripts
- Gene Symbol
- MFSD4A
- Entrez Gene ID
- 148808
- Gene Name
- major facilitator superfamily domain containing 4A
- Gene Aliases
- MFSD4, UNQ3064
- Chromosome Location
- Chr. 1: 205568984 - 205602918 on Build GRCh38
- UniGene ID
- Hs.567714
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_181644.4 |
NP_857595.3
|
9, 10 |
1571 |
|
XM_011509229.2 |
XP_011507531.1
|
8, 9 |
1310 |
|
GenBank mRNA |
AK057733.1 |
- |
5, 6 |
641 |
|
AK090831.1 |
- |
3, 4 |
298 |
|
AK091896.1 |
- |
9, 10 |
1512 |
|
AK127321.1 |
- |
1, 2 |
1207 |
|
AK293816.1 |
- |
8, 9 |
1500 |
|
AK295148.1 |
- |
8, 9 |
1158 |
|
AK304109.1 |
- |
8, 9 |
1376 |
|
AK304120.1 |
- |
8, 9 |
1237 |
|
AL713676.1 |
- |
7, 8 |
877 |
|
AY358107.1 |
- |
7, 8 |
958 |
|
BC036549.2 |
- |
9, 10 |
1521 |
5264261 |
View Full Product Details