Gene Transcripts
- Gene Symbol
- CNIH3
- Entrez Gene ID
- 149111
- Gene Name
- cornichon family AMPA receptor auxiliary protein 3
- Gene Aliases
- CNIH-3
- Chromosome Location
- Chr. 1: 224434640 - 224740549 on Build GRCh38
- UniGene ID
- Hs.731723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001322302.1 |
NP_001309231.1
|
3, 4 |
435 |
|
NM_001322303.1 |
NP_001309232.1
|
2, 3 |
369 |
|
NM_001322304.1 |
NP_001309233.1
|
3, 4 |
513 |
|
NM_152495.1 |
NP_689708.1
|
2, 3 |
855 |
|
NR_136287.1 |
|
2, 3 |
855 |
|
NR_136288.1 |
|
3, 4 |
365 |
|
NR_136289.1 |
|
4, 5 |
501 |
|
NR_136290.1 |
|
3, 4 |
435 |
|
NR_136291.1 |
|
4, 5 |
571 |
|
NR_136292.1 |
|
3, 4 |
505 |
|
NR_136293.1 |
|
6, 7 |
741 |
|
NR_136294.1 |
|
5, 6 |
675 |
|
NR_136295.1 |
|
4, 5 |
546 |
|
NR_136296.1 |
|
4, 5 |
571 |
|
NR_136297.1 |
|
3, 4 |
435 |
|
GenBank mRNA |
AF070524.1 |
- |
1, 2 |
61 |
|
AK096312.1 |
- |
2, 3 |
855 |
|
BC022780.1 |
- |
2, 3 |
343 |
3894832 |
View Full Product Details