Gene Transcripts

Gene Symbol
DNAJB7
Entrez Gene ID
150353
Gene Name
DnaJ heat shock protein family (Hsp40) member B7
Gene Aliases
DJ5, HSC3
Chromosome Location
Chr. 22: 40859549 - 40862126 on Build GRCh38
UniGene ID
Hs.585042
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145174.1 NP_660157.1 1 994
GenBank mRNA AF085232.1 - 1 994
BC112135.1 - 1 953 8327603
CR456468.1 - 1 899

View Full Product Details