Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001967.3 | NP_001958.2 | 11 | 1212 | |
GenBank mRNA | AB209021.1 | - | 3 | 4591 | |
AF208852.1 | - | 10 | 1050 | ||
AK130087.1 | - | 11 | 1207 | ||
AK225646.1 | - | 11 | 1207 | ||
AL117412.1 | - | 10 | 1072 | ||
AL137681.1 | - | 4 | 3238 | ||
BC012547.1 | - | 11 | 1214 | 4079371 | |
BC013708.1 | - | 11 | 1192 | 4341420 | |
BC015842.1 | - | 11 | 1211 | 4337581 | |
BC016295.2 | - | 5 | 490 | ||
BC048105.1 | - | 11 | 1210 | 4820774 | |
BT009860.1 | - | 11 | 1173 | ||
D30655.1 | - | 11 | 1188 | ||
S79942.1 | - | 11 | 1188 |