Gene Transcripts

Gene Symbol
OST4
Entrez Gene ID
100128731
Gene Name
oligosaccharyltransferase complex subunit 4, non-catalytic
Gene Aliases
Chromosome Location
Chr. 2: 27070472 - 27071699 on Build GRCh38
UniGene ID
Hs.502948
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134693.1 NP_001128165.1 3 340
GenBank mRNA AL049447.1 - 3 266
BC105283.1 - 3 298
BF697310.1 - 3 341

View Full Product Details