Gene Transcripts

Gene Symbol
SPINT3
Entrez Gene ID
10816
Gene Name
serine peptidase inhibitor, Kunitz type 3
Gene Aliases
HKIB9
Chromosome Location
Chr. 20: 45512461 - 45515624 on Build GRCh38
UniGene ID
Hs.370724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006652.1 NP_006643.1 2 111
GenBank mRNA AY372172.2 - 2 111
BX088813.1 - 2 61

View Full Product Details