Gene Transcripts
- Gene Symbol
- OR1Q1
- Entrez Gene ID
- 158131
- Gene Name
- olfactory receptor family 1 subfamily Q member 1
- Gene Aliases
- HSTPCR106, OR1Q2, OR1Q3, OR9-25, OR9-A, OST226, OST226OR9-A, TPCR106
- Chromosome Location
- Chr. 9: 122614738 - 122615682 on Build GRCh38
- UniGene ID
- Hs.684864
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012364.1 |
NP_036496.1
|
1 |
479 |
|
View Full Product Details