Gene Transcripts

Gene Symbol
CLECL1
Entrez Gene ID
160365
Gene Name
C-type lectin like 1
Gene Aliases
DCAL-1, DCAL1
Chromosome Location
Chr. 12: 9715860 - 9733435 on Build GRCh38
UniGene ID
Hs.560087
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253750.1 NP_001240679.1 1 94
NM_001267701.1 NP_001254630.1 1 94
NM_172004.3 NP_742001.1 1 94
XM_011520574.2 XP_011518876.1 1 137
GenBank mRNA AF518873.1 - 1 59
BC093857.1 - 1 94 7939702
BC093859.1 - 1 94 7939704

View Full Product Details