Gene Transcripts

Gene Symbol
ANKFN1
Entrez Gene ID
162282
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases
Chromosome Location
Chr. 17: 56110935 - 56517016 on Build GRCh38
UniGene ID
Hs.673040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153228.2 NP_694960.2 10, 11 1243
XM_006721728.3 XP_006721791.1 9, 10 1466
XM_011524428.2 XP_011522730.1 10, 11 1243
XM_011524429.2 XP_011522731.1 10, 11 1400
XM_011524430.2 XP_011522732.1 11, 12 1366
XM_011524431.2 XP_011522733.1 10, 11 1298
XM_011524433.2 XP_011522735.1 4, 5 656
XM_017024263.1 XP_016879752.1 12, 13 1460
XM_017024264.1 XP_016879753.1 12, 13 2445
XM_017024265.1 XP_016879754.1 12, 13 1654
XM_017024266.1 XP_016879755.1 10, 11 1415
XM_017024267.1 XP_016879756.1 9, 10 1267
XM_017024268.1 XP_016879757.1 11, 12 2351
XM_017024269.1 XP_016879758.1 9, 10 1540
XM_017024270.1 XP_016879759.1 12, 13 1458
GenBank mRNA AK095654.1 - 10, 11 1243

View Full Product Details