Gene Transcripts

Gene Symbol
EMP2
Entrez Gene ID
2013
Gene Name
epithelial membrane protein 2
Gene Aliases
XMP
Chromosome Location
Chr. 16: 10528422 - 10580716 on Build GRCh38
UniGene ID
Hs.531561
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001424.5 NP_001415.1 3 365
XM_006720864.3 XP_006720927.1 3 359
GenBank mRNA AK313134.1 - 3 281
AV657588.1 - 2 80
BC009687.1 - 3 256 3895094
BF981157.1 - 3 347
HY116508.1 - 3 366
U52100.1 - 2 200
X94770.1 - 2 147

View Full Product Details