Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170574.1 | NP_001164045.1 | 5 | 1090 | |
NM_152630.4 | NP_689843.1 | 3 | 1014 | ||
GenBank mRNA | AK128147.1 | - | 5 | 1090 | |
AK313875.1 | - | 3 | 985 | ||
BC028710.2 | - | 3 | 983 | 4827590 | |
BC030784.2 | - | 3 | 995 | 4826421 | |
BC034979.2 | - | 3 | 980 | 4824369 | |
BX537938.1 | - | 3 | 1016 |