Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181715.2 | NP_859066.1 | 13 | 2007 | |
XM_005244946.1 | XP_005245003.1 | 13 | 2003 | ||
XM_005244949.2 | XP_005245006.1 | 13 | 1880 | ||
XM_017000575.1 | XP_016856064.1 | 12 | 1776 | ||
XM_017000576.1 | XP_016856065.1 | 12 | 1761 | ||
XM_017000577.1 | XP_016856066.1 | 12 | 1905 | ||
XM_017000578.1 | XP_016856067.1 | 12 | 1890 | ||
XM_017000579.1 | XP_016856068.1 | 7 | 1302 | ||
GenBank mRNA | AF289574.1 | - | 4 | 966 | |
AK096382.1 | - | 11 | 1598 | ||
AK223559.1 | - | 13 | 1960 | ||
AY360172.1 | - | 13 | 1978 | ||
AY726597.1 | - | 13 | 1787 | ||
BC028886.1 | - | 2 | 242 | ||
BC033304.1 | - | 12 | 1676 | ||
BC053562.1 | - | 13 | 1918 | 6188068 |