Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_181715.2 | NP_859066.1 | 4 | 554 | |
XM_005244946.1 | XP_005245003.1 | 4 | 535 | ||
XM_005244949.2 | XP_005245006.1 | 4 | 412 | ||
XM_017000575.1 | XP_016856064.1 | 4 | 412 | ||
XM_017000576.1 | XP_016856065.1 | 4 | 412 | ||
XM_017000577.1 | XP_016856066.1 | 4 | 541 | ||
XM_017000578.1 | XP_016856067.1 | 4 | 541 | ||
GenBank mRNA | AK096382.1 | - | 3 | 537 | |
AK223559.1 | - | 4 | 507 | ||
AY360172.1 | - | 4 | 525 | ||
AY726597.1 | - | 4 | 334 | ||
BC033304.1 | - | 2 | 141 | ||
BC053562.1 | - | 4 | 465 | 6188068 | |
BI550818.1 | - | 4 | 561 |