Gene Transcripts

Gene Symbol
FAM219A
Entrez Gene ID
203259
Gene Name
family with sequence similarity 219 member A
Gene Aliases
C9orf25
Chromosome Location
Chr. 9: 34398184 - 34458610 on Build GRCh38
UniGene ID
Hs.493771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184940.1 NP_001171869.1 5 668
NM_001184941.1 NP_001171870.1 5 668
NM_001184942.1 NP_001171871.1 5 635
NM_001184943.1 NP_001171872.1 5 617
NM_001184945.1 NP_001171874.1 5 584
NM_147202.1 NP_671735.1 5 617
XM_005251402.4 XP_005251459.1 6 875
XM_011517797.2 XP_011516099.1 5 675
XM_011517798.2 XP_011516100.1 5 624
XM_011517799.2 XP_011516101.1 5 473
GenBank mRNA AK096350.1 - 5 617
AK294054.1 - 5 635
AK299882.1 - 5 663
BC025348.1 - 1 25
BC041009.2 - 5 584 5284905
DA110626.1 - 5 407

View Full Product Details