Gene Transcripts

Gene Symbol
C3orf58
Entrez Gene ID
205428
Gene Name
chromosome 3 open reading frame 58
Gene Aliases
DIA1, GoPro49, HASF
Chromosome Location
Chr. 3: 143971798 - 143992368 on Build GRCh38
UniGene ID
Hs.288954
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134470.1 NP_001127942.1 2 488
NM_173552.3 NP_775823.1 2 1442
XM_017005869.1 XP_016861358.1 2 322
GenBank mRNA AK095161.1 - 2 504
AK095400.1 - 2 348
AK294020.1 - 2 386
AK315082.1 - 2 1416
AL832839.1 - 2 504
BC037293.2 - 2 1169 5267770
DC310421.1 - 2 386

View Full Product Details