Gene Transcripts

Gene Symbol
OOSP2
Entrez Gene ID
219990
Gene Name
oocyte secreted protein 2
Gene Aliases
PLAC1L, TMEM122
Chromosome Location
Chr. 11: 60040275 - 60048044 on Build GRCh38
UniGene ID
Hs.132310
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173801.4 NP_776162.2 3 464
GenBank mRNA AK093517.1 - 3 330
BC036256.1 - 3 330 5270384
BC048121.1 - 3 464 5266642

View Full Product Details