Gene Transcripts

Gene Symbol
RNASEH1
Entrez Gene ID
246243
Gene Name
ribonuclease H1
Gene Aliases
H1RNA, PEOB2, RNH1
Chromosome Location
Chr. 2: 3531813 - 3558371 on Build GRCh38
UniGene ID
Hs.568006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286834.1 NP_001273763.1 8 913
NM_001286837.1 NP_001273766.1 8 1151
NM_002936.4 NP_002927.2 8 913
GenBank mRNA AF039652.1 - 8 883
AF048994.1 - 8 829
AF048995.1 - 8 863
AJ224117.1 - 8 802
AK075490.1 - 8 862
AK096913.1 - 8 1069
BC002973.1 - 8 872 3537074
BT019670.1 - 8 802
CR541820.1 - 8 802

View Full Product Details