Gene Transcripts

Gene Symbol
OR1L4
Entrez Gene ID
254973
Gene Name
olfactory receptor family 1 subfamily L member 4
Gene Aliases
OR1L5, OR9-29, OR9-E, OST046
Chromosome Location
Chr. 9: 122723990 - 122724925 on Build GRCh38
UniGene ID
Hs.626838
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005235.1 NP_001005235.1 1 403

Gene Symbol
OR1L6
Entrez Gene ID
392390
Gene Name
olfactory receptor family 1 subfamily L member 6
Gene Aliases
HG16, OR1K1, OR1L7, OR9-30
Chromosome Location
Chr. 9: 122749848 - 122750783 on Build GRCh38
UniGene ID
Hs.553790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004453.2 NP_001004453.2 1 403

View Full Product Details