Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001030019.1 | NP_001025190.1 | 5 | 592 | |
NM_001284350.1 | NP_001271279.1 | 6 | 690 | ||
NM_152782.3 | NP_689995.3 | 6 | 508 | ||
XM_017011930.1 | XP_016867419.1 | 6 | 498 | ||
GenBank mRNA | AF429967.1 | - | 5 | 592 | |
AK302011.1 | - | 6 | 619 | ||
BC026189.2 | - | 6 | 496 | 4819018 | |
BG772576.1 | - | 6 | 513 | ||
BI825617.1 | - | 5 | 571 | ||
DQ099386.1 | - | 2 | 111 |