Gene Transcripts

Gene Symbol
F3
Entrez Gene ID
2152
Gene Name
coagulation factor III, tissue factor
Gene Aliases
CD142, TF, TFA
Chromosome Location
Chr. 1: 94529176 - 94541857 on Build GRCh38
UniGene ID
Hs.62192
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178096.1 NP_001171567.1 2 422
NM_001993.4 NP_001984.1 2 422
GenBank mRNA AF487337.1 - 2 202
AF497569.1 - 2 149
AF497570.1 - 2 149
AK310918.1 - 3 816
AK312638.1 - 2 318
AY940729.1 - 3 819
AY940730.1 - 2 202
AY940731.1 - 2 202
BC011029.1 - 2 369 4073183
BT019808.1 - 2 201
CR541792.1 - 2 201
DB203987.1 - 2 380
DB499288.1 - 2 424
J02931.1 - 2 312
M16553.1 - 2 276
M27436.1 - 2 300

View Full Product Details